Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSGALNACT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261473.1 | NP_001248402.1 | 947 | 91 | ||
XM_015144797.2 | XP_015000283.1 | 1306 | 91 | |||
XM_015144799.2 | XP_015000285.1 | 1355 | 91 | |||
XM_015144802.2 | XP_015000288.1 | 1473 | 91 | |||
XM_028852233.1 | XP_028708066.1 | 1464 | 91 | |||
XM_028852235.1 | XP_028708068.1 | 1412 | 91 | |||
GenBank mRNA | JU322583.1 | - | 947 | 91 | ||
JV474384.1 | - | 246 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |