Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRCIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082491.4 | XP_001082491.4 | 2275 | 98 | ||
XM_015119493.2 | XP_014974979.2 | 2439 | 98 | |||
XM_015119494.2 | XP_014974980.1 | 3863 | 98 | |||
XM_015119500.2 | XP_014974986.1 | 1973 | 98 | |||
XM_028836428.1 | XP_028692261.1 | 2241 | 98 | |||
XM_028836429.1 | XP_028692262.1 | 2449 | 98 | |||
XM_028836430.1 | XP_028692263.1 | 2438 | 98 | |||
XM_028836431.1 | XP_028692264.1 | 2336 | 98 | |||
GenBank mRNA | JU348927.1 | - | 1927 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |