Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGPEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260688.1 | NP_001247617.1 | 288 | 63 | ||
XM_015123307.2 | XP_014978793.1 | 406 | 63 | |||
XM_015123309.2 | XP_014978795.1 | 316 | 63 | |||
XM_028838550.1 | XP_028694383.1 | 301 | 63 | |||
XM_028838551.1 | XP_028694384.1 | 316 | 63 | |||
GenBank mRNA | JU473664.1 | - | 288 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |