Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126470.2 | XP_014981956.1 | 422 | 70 | ||
XM_015126472.2 | XP_014981958.1 | 387 | 70 | |||
XM_015126473.2 | XP_014981959.1 | 447 | 70 | |||
XM_015126474.2 | XP_014981960.1 | 395 | 70 | |||
XM_015126475.2 | XP_014981961.1 | 337 | 70 | |||
XM_015126476.2 | XP_014981962.1 | 255 | 70 | |||
XM_015126477.2 | XP_014981963.1 | 354 | 70 | |||
XM_015126478.2 | XP_014981964.1 | 270 | 70 | |||
GenBank mRNA | JU322849.1 | - | 181 | 70 | ||
JU322850.1 | - | 181 | 70 | |||
JU472707.1 | - | 161 | 70 | |||
JU472708.1 | - | 161 | 70 | |||
JV046253.1 | - | 188 | 70 | |||
JV046254.1 | - | 188 | 70 | |||
JV707539.1 | - | 195 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |