Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015114859.2 | XP_014970345.1 | 707 | 99 | ||
XM_015114860.2 | XP_014970346.1 | 707 | 99 | |||
XM_015114861.2 | XP_014970347.1 | 658 | 99 | |||
GenBank mRNA | JU321825.1 | - | 631 | 99 | ||
JV046995.1 | - | 399 | 99 | |||
JV601073.1 | - | 655 | 99 | |||
JV668030.1 | - | 652 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |