Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028828738.1 | XP_028684571.1 | 1085 | 134 | ||
XM_028828739.1 | XP_028684572.1 | 1146 | 134 | |||
XM_028828740.1 | XP_028684573.1 | 1127 | 134 | |||
XM_028828741.1 | XP_028684574.1 | 1088 | 134 | |||
XM_028828742.1 | XP_028684575.1 | 993 | 134 | |||
GenBank mRNA | JV606404.1 | - | 979 | 134 | ||
JV642761.1 | - | 971 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |