Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028828738.1 | XP_028684571.1 | 1364 | 76 | ||
XM_028828739.1 | XP_028684572.1 | 1425 | 76 | |||
XM_028828740.1 | XP_028684573.1 | 1406 | 76 | |||
XM_028828741.1 | XP_028684574.1 | 1367 | 76 | |||
XM_028828742.1 | XP_028684575.1 | 1272 | 76 | |||
GenBank mRNA | JV606404.1 | - | 1258 | 76 | ||
JV642761.1 | - | 1250 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |