Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015134306.2 | XP_014989792.1 | 855 | 90 | ||
GenBank mRNA | JV708570.1 | - | 196 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |