Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083289.4 | XP_001083289.1 | 623 | 119 | ||
XM_015145669.2 | XP_015001155.1 | 620 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |