Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GALNAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261629.1 | NP_001248558.1 | 892 | 102 | ||
GenBank mRNA | JU333826.1 | - | 892 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |