Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3GL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001111376.4 | XP_001111376.2 | 739 | 122 | ||
XM_015142897.2 | XP_014998383.1 | 732 | 122 | |||
XM_028851244.1 | XP_028707077.1 | 817 | 122 | |||
GenBank mRNA | JU326923.1 | - | 210 | 122 | ||
JV590849.1 | - | 444 | 122 | |||
JV625851.1 | - | 425 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |