Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086701.4 | XP_001086701.4 | 1017 | 65 | ||
GenBank mRNA | JV046358.1 | - | 779 | 65 | ||
JV636003.1 | - | 806 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |