Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001109737.4 | XP_001109737.1 | 1445 | 66 | ||
XM_015132140.2 | XP_014987626.1 | 1654 | 66 | |||
XM_015132149.2 | XP_014987635.1 | 1385 | 66 | |||
XM_015132157.2 | XP_014987643.1 | 1875 | 66 | |||
XM_015132161.2 | XP_014987647.1 | 2332 | 66 | |||
XM_015132170.2 | XP_014987656.1 | 2149 | 66 | |||
XM_015132179.2 | XP_014987665.1 | 1536 | 66 | |||
XM_028837504.1 | XP_028693337.1 | 2089 | 66 | |||
GenBank mRNA | JU322912.1 | - | 1248 | 66 | ||
JU472639.1 | - | 1263 | 66 | |||
JV043498.1 | - | 1206 | 66 | |||
JV639321.1 | - | 1321 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |