Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUNDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032900.1 | NP_001028072.1 | 361 | 68 | ||
GenBank mRNA | AY340968.1 | - | 361 | 68 | ||
JV601141.1 | - | 465 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |