Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257614.1 | NP_001244543.1 | 1305 | 60 | ||
XM_015127466.1 | XP_014982952.1 | 1432 | 60 | |||
XM_015127467.2 | XP_014982953.1 | 1353 | 60 | |||
XM_015127468.2 | XP_014982954.1 | 1378 | 60 | |||
XM_015127469.2 | XP_014982955.1 | 1429 | 60 | |||
GenBank mRNA | JU321428.1 | - | 1375 | 60 | ||
JU321429.1 | - | 1305 | 60 | |||
JU471522.1 | - | 1366 | 60 | |||
JU471523.1 | - | 1305 | 60 | |||
JV528057.1 | - | 1306 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |