Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUPT4H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260659.1 | NP_001247588.1 | 340 | 74 | ||
GenBank mRNA | JU336098.1 | - | 357 | 74 | ||
JU472450.1 | - | 362 | 74 | |||
JV044853.1 | - | 340 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |