Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260936.1 | NP_001247865.1 | 304 | 56 | ||
XM_028832693.1 | XP_028688526.1 | 904 | 56 | |||
GenBank mRNA | JU334464.1 | - | 295 | 56 | ||
JU334465.1 | - | 295 | 56 | |||
JU475552.1 | - | 304 | 56 | |||
JU475553.1 | - | 304 | 56 | |||
JU475554.1 | - | 304 | 56 | |||
JU475555.1 | - | 304 | 56 | |||
JU475556.1 | - | 304 | 56 | |||
JU475557.1 | - | 304 | 56 | |||
JU475558.1 | - | 304 | 56 | |||
JU475559.1 | - | 304 | 56 | |||
JV565116.1 | - | 300 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |