Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260885.1 | NP_001247814.1 | 86 | 77 | ||
XM_015138318.2 | XP_014993804.1 | 143 | 77 | |||
GenBank mRNA | JU476157.1 | - | 86 | 77 |