Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115200.2 | XP_014970686.1 | 376 | 64 | ||
XM_028833816.1 | XP_028689649.1 | 348 | 64 | |||
XM_028833817.1 | XP_028689650.1 | 550 | 64 | |||
XM_028833818.1 | XP_028689651.1 | 162 | 64 | |||
GenBank mRNA | JU324173.1 | - | 331 | 64 | ||
JU473343.1 | - | 317 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |