Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP26A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193915.2 | NP_001180844.1 | 961 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |