Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPCS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194879.3 | NP_001181808.1 | 197 | 93 | ||
GenBank mRNA | JU320315.1 | - | 265 | 93 | ||
JU476682.1 | - | 163 | 93 | |||
JV047855.1 | - | 258 | 93 | |||
JV600883.1 | - | 282 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |