Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193873.1 | NP_001180802.1 | 401 | 121 | ||
XM_015110976.1 | XP_014966462.1 | 396 | 121 | |||
GenBank mRNA | JU335713.1 | - | 160 | 121 | ||
JU476597.1 | - | 158 | 121 | |||
JV047795.1 | - | 141 | 121 | |||
JV589402.1 | - | 132 | 121 | |||
JV600818.1 | - | 149 | 121 | |||
JV667440.1 | - | 145 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |