Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015123285.2 | XP_014978771.2 | 673 | 93 | ||
XM_015123286.2 | XP_014978772.1 | 758 | 93 | |||
XM_015123287.2 | XP_014978773.2 | 673 | 93 | |||
XM_028838956.1 | XP_028694789.1 | 673 | 93 | |||
XM_028838957.1 | XP_028694790.1 | 673 | 93 | |||
XM_028838958.1 | XP_028694791.1 | 745 | 93 | |||
XM_028838959.1 | XP_028694792.1 | 745 | 93 | |||
GenBank mRNA | JU334427.1 | - | 490 | 93 | ||
JU334428.1 | - | 490 | 93 | |||
JV566310.1 | - | 492 | 93 | |||
JV581195.1 | - | 492 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |