Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEZF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028844309.1 | XP_028700142.1 | 622 | 113 | ||
GenBank mRNA | JV620795.1 | - | 211 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |