Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194106.1 | NP_001181035.1 | 345 | 69 | ||
XM_015127705.2 | XP_014983191.1 | 420 | 69 | |||
GenBank mRNA | JU322812.1 | - | 245 | 69 | ||
JU475866.1 | - | 245 | 69 | |||
JV508990.1 | - | 248 | 69 | |||
JV653154.1 | - | 267 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |