Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260560.1 | NP_001247489.1 | 341 | 100 | ||
GenBank mRNA | JU327851.1 | - | 269 | 100 | ||
JV044968.1 | - | 341 | 100 | |||
JV635708.1 | - | 592 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |