Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257826.1 | NP_001244755.1 | 2143 | 68 | ||
XM_028840352.1 | XP_028696185.1 | 1745 | 68 | |||
GenBank mRNA | JU322627.1 | - | 2143 | 68 | ||
JV723736.1 | - | 2133 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |