Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257923.1 | NP_001244852.1 | 293 | 56 | ||
XM_028846419.1 | XP_028702252.1 | 286 | 56 | |||
GenBank mRNA | JU332259.1 | - | 293 | 56 | ||
JU473726.1 | - | 258 | 56 | |||
JV044736.1 | - | 282 | 56 | |||
JV565151.1 | - | 213 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |