Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266627.1 | NP_001253556.1 | 512 | 75 | ||
XM_015123572.2 | XP_014979058.1 | 134 | 75 | |||
GenBank mRNA | JV043228.1 | - | 512 | 75 |
Gene Symbol: TBCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU319753.1 | - | 376 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |