Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257302.1 | NP_001244231.1 | 111 | 94 | ||
NM_001271648.2 | NP_001258577.2 | 111 | 94 | |||
GenBank mRNA | JQ038001.2 | - | 111 | 94 | ||
JQ038002.2 | - | 111 | 94 | |||
JQ038003.1 | - | 111 | 94 | |||
JU482521.1 | - | 127 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |