Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HACD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261129.1 | NP_001248058.1 | 391 | 105 | ||
XM_028843389.1 | XP_028699222.1 | 417 | 105 | |||
XM_028843390.1 | XP_028699223.1 | 195 | 105 | |||
XM_028843391.1 | XP_028699224.1 | 438 | 105 | |||
XM_028843392.1 | XP_028699225.1 | 101 | 105 | |||
GenBank mRNA | JU327265.1 | - | 366 | 105 | ||
JV044709.1 | - | 391 | 105 | |||
JV639352.1 | - | 387 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |