Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032829.1 | NP_001028001.1 | 738 | 92 | ||
GenBank mRNA | AF190980.1 | - | 375 | 92 | ||
AF294891.1 | - | 738 | 92 | |||
AF395618.1 | - | 375 | 92 | |||
AJ585529.1 | - | 354 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |