Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032820.2 | NP_001027992.1 | 104 | 128 | ||
GenBank mRNA | AF240769.1 | - | 98 | 128 | ||
JV044681.1 | - | 91 | 128 | |||
JV565238.1 | - | 103 | 128 | |||
JV635626.1 | - | 106 | 128 | |||
JV669158.1 | - | 104 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |