Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032831.1 | NP_001028003.1 | 99 | 62 | ||
GenBank mRNA | AF303085.1 | - | 99 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |