Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110262.1 | NP_001103732.1 | 299 | 103 | ||
GenBank mRNA | AF466354.1 | - | 299 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |