Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015124856.2 | XP_014980342.2 | 500 | 69 | ||
GenBank mRNA | AY386685.1 | - | 355 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |