Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2A24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040215.1 | NP_001035305.1 | 487 | 90 | ||
XM_028838334.1 | XP_028694167.1 | 523 | 90 | |||
GenBank mRNA | AY635460.1 | - | 487 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |