Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195492.1 | NP_001182421.1 | 87 | 125 | ||
GenBank mRNA | DQ147992.1 | - | 83 | 126 | ||
DV769372.2 | - | 111 | 125 | |||
JU319669.1 | - | 202 | 125 | |||
JU319670.1 | - | 202 | 125 | |||
JU319671.1 | - | 202 | 125 | |||
JU473502.1 | - | 136 | 125 | |||
JU473503.1 | - | 136 | 125 | |||
JV045016.1 | - | 131 | 125 | |||
JV045017.1 | - | 131 | 125 | |||
JV601008.1 | - | 218 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |