Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6AP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257625.1 | NP_001244554.1 | 966 | 144 | ||
GenBank mRNA | JU332431.1 | - | 966 | 144 | ||
JU473893.1 | - | 892 | 144 | |||
JV045853.1 | - | 918 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |