Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROPN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015139739.2 | XP_014995225.2 | 1635 | 117 | ||
XM_015139740.2 | XP_014995226.2 | 1358 | 117 | |||
XM_028849340.1 | XP_028705173.1 | 551 | 117 | |||
GenBank mRNA | JV595465.1 | - | 137 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |