Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257879.1 | NP_001244808.1 | 297 | 74 | ||
GenBank mRNA | JU335241.1 | - | 297 | 74 | ||
JU476211.1 | - | 304 | 74 | |||
JV047507.1 | - | 308 | 74 | |||
JV655035.1 | - | 322 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |