Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS2ST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260915.1 | NP_001247844.1 | 818 | 153 | ||
GenBank mRNA | JU337259.1 | - | 523 | 153 | ||
JU477966.1 | - | 498 | 153 | |||
JV048806.1 | - | 818 | 153 | |||
JV634365.1 | - | 838 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |