Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORMDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266349.1 | NP_001253278.1 | 436 | 138 | ||
XM_015110498.2 | XP_014965984.1 | 1656 | 138 | |||
XM_015110499.2 | XP_014965985.1 | 558 | 138 | |||
XM_015110500.2 | XP_014965986.1 | 466 | 138 | |||
XM_028830555.1 | XP_028686388.1 | 616 | 138 | |||
XM_028830556.1 | XP_028686389.1 | 585 | 138 | |||
GenBank mRNA | JU320839.1 | - | 435 | 138 | ||
JU477898.1 | - | 436 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |