Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLOC1S6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001111943.4 | XP_001111943.1 | 627 | 137 | ||
XM_015142148.2 | XP_014997634.1 | 627 | 137 | |||
XM_028850880.1 | XP_028706713.1 | 627 | 137 | |||
GenBank mRNA | JV703971.1 | - | 516 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |