Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194343.2 | NP_001181272.1 | 701 | 71 | ||
GenBank mRNA | JU323207.1 | - | 747 | 71 | ||
JU471549.1 | - | 682 | 71 | |||
JV045400.1 | - | 671 | 71 | |||
JV600534.1 | - | 775 | 71 | |||
JV637676.1 | - | 719 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |