Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPSM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257808.2 | NP_001244737.1 | 42 | 85 | ||
GenBank mRNA | JU335869.1 | - | 42 | 85 | ||
JU476747.1 | - | 81 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |