Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082488.4 | XP_001082488.1 | 575 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |