Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266445.1 | NP_001253374.1 | 389 | 158 | ||
XM_015109893.1 | XP_014965379.1 | 330 | 158 | |||
GenBank mRNA | JU332513.1 | - | 440 | 158 | ||
JU332514.1 | - | 296 | 158 | |||
JU473988.1 | - | 407 | 158 | |||
JU473989.1 | - | 263 | 158 | |||
JV045914.1 | - | 389 | 158 | |||
JV705243.1 | - | 462 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |