Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRFN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015136304.2 | XP_014991790.2 | 2189 | 107 | ||
GenBank mRNA | JV588470.1 | - | 1401 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |