Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015117824.2 | XP_014973310.2 | 1155 | 103 | ||
XM_015117825.2 | XP_014973311.2 | 1108 | 103 | |||
GenBank mRNA | JV053672.1 | - | 901 | 103 | ||
JV635996.1 | - | 1006 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |